Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGCTGTAAAAATCCAAGCAAGA[C/T]GGACTACATAGAGTTAAAATTTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608065 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC38A4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC38A4 - solute carrier family 38 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143824.1 | Intron | NP_001137296.1 | ||||
NM_018018.4 | Intron | NP_060488.2 | ||||
XM_005268997.2 | Intron | XP_005269054.1 |