Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGGCCTGTGTTTCTGGGACCTA[A/C]GGAGGCAAGCATAACTTTACATCAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
TMEM44 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
TMEM44 - transmembrane protein 44 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011655.2 | Intron | NP_001011655.1 | ||||
NM_001166305.1 | Intron | NP_001159777.1 | ||||
NM_001166306.1 | Intron | NP_001159778.1 | ||||
NM_138399.4 | Intron | NP_612408.3 | ||||
XM_005269371.4 | Intron | XP_005269428.1 | ||||
XM_011513318.2 | Intron | XP_011511620.1 | ||||
XM_011513319.2 | Intron | XP_011511621.1 | ||||
XM_011513320.2 | Intron | XP_011511622.1 | ||||
XM_011513321.2 | Intron | XP_011511623.1 | ||||
XM_011513322.2 | Intron | XP_011511624.1 | ||||
XM_017007517.1 | Intron | XP_016863006.1 | ||||
XM_017007518.1 | Intron | XP_016863007.1 |
TMEM44-AS1 - TMEM44 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |