Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTGCTCCCCACTCCAGGCCCAC[A/G]CACACCTACCTGTGAAGTACCTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604090 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DLG5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DLG5 - discs large MAGUK scaffold protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004747.3 | Intron | NP_004738.3 | ||||
XM_005270276.4 | Intron | XP_005270333.1 | ||||
XM_006718056.3 | Intron | XP_006718119.1 | ||||
XM_006718057.3 | Intron | XP_006718120.1 | ||||
XM_011540341.2 | Intron | XP_011538643.1 | ||||
XM_011540342.1 | Intron | XP_011538644.1 | ||||
XM_011540344.2 | Intron | XP_011538646.1 | ||||
XM_011540345.1 | Intron | XP_011538647.1 | ||||
XM_011540346.2 | Intron | XP_011538648.1 | ||||
XM_011540347.2 | Intron | XP_011538649.1 | ||||
XM_017016913.1 | Intron | XP_016872402.1 | ||||
XM_017016914.1 | Intron | XP_016872403.1 | ||||
XM_017016915.1 | Intron | XP_016872404.1 |