Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAACAAAACCAAACAAAAAACCC[C/T]AGCAACAACAAGATATTCTCTTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTBD16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BTBD16 - BTB domain containing 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318189.1 | 1138 | Intron | NP_001305118.1 | |||
NM_144587.3 | 1138 | Intron | NP_653188.2 | |||
XM_011539239.2 | 1138 | Intron | XP_011537541.1 | |||
XM_011539240.2 | 1138 | Intron | XP_011537542.1 | |||
XM_011539241.2 | 1138 | Intron | XP_011537543.1 | |||
XM_011539242.2 | 1138 | Intron | XP_011537544.1 | |||
XM_017015636.1 | 1138 | UTR 5 | XP_016871125.1 | |||
XM_017015637.1 | 1138 | UTR 5 | XP_016871126.1 |