Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGGTTGGGGACCCCTGCTTTAA[A/G]GGAAGGGACTTACTCTTAGTAATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607657 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTH - cystathionine gamma-lyase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190463.1 | Intron | NP_001177392.1 | ||||
NM_001902.5 | Intron | NP_001893.2 | ||||
NM_153742.4 | Intron | NP_714964.2 | ||||
XM_005270509.3 | Intron | XP_005270566.1 | ||||
XM_017000416.1 | Intron | XP_016855905.1 |