Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTTCCTTGTAGAGGGAACAGCCA[A/G]TGTGAAAGCACAGAGTCAGGATCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601741 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CUL5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CUL5 - cullin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003478.3 | Intron | NP_003469.2 | ||||
XM_005271682.2 | Intron | XP_005271739.1 | ||||
XM_011543013.1 | Intron | XP_011541315.1 | ||||
XM_017018363.1 | Intron | XP_016873852.1 | ||||
XM_017018364.1 | Intron | XP_016873853.1 | ||||
XM_017018365.1 | Intron | XP_016873854.1 |