Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCTCAGCCTCGTGGGCTCGGCGG[A/G]CTGCAGCGAGAAGCACCTGCTGGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 152390 | ||||||||||||||||||||
Literature Links: |
ALOX5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALOX5 - arachidonate 5-lipoxygenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000698.4 | 206 | Missense Mutation | GAC,GGC | D,G 31 | NP_000689.1 | |
NM_001256153.2 | 206 | Missense Mutation | GAC,GGC | D,G 31 | NP_001243082.1 | |
NM_001256154.2 | 206 | Missense Mutation | GAC,GGC | D,G 31 | NP_001243083.1 | |
NM_001320861.1 | 206 | Missense Mutation | GAC,GGC | D,G 31 | NP_001307790.1 | |
NM_001320862.1 | 206 | UTR 5 | NP_001307791.1 | |||
XM_017016012.1 | 206 | UTR 5 | XP_016871501.1 |