Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTAAGTTGGTGGCAGGAACCAGC[C/T]TCCTGAAGGATATAAATTCGCTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601054 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLXNA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLXNA2 - plexin A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025179.3 | 10210 | UTR 3 | NP_079455.3 | |||
XM_005273164.2 | 10210 | UTR 3 | XP_005273221.1 | |||
XM_005273165.3 | 10210 | Intron | XP_005273222.1 |