Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACGGTGCTGGTGGTCTTCCGCAT[C/T]GTGCTGACGGCTGTGGGCGGCGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608803 MIM: 139270 MIM: 615316 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GJC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GJC2 - gap junction protein gamma 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020435.3 | 421 | Silent Mutation | ATC,ATT | I,I 36 | NP_065168.2 |
GUK1 - guanylate kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IBA57 - IBA57 homolog, iron-sulfur cluster assembly | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IBA57-AS1 - IBA57 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |