Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTTCATCTATCTAGATCGCCTCT[C/T]CCGCTGCCGCTTCAACACCTCCAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608554 | ||||||||||||||||||||
Literature Links: |
NPAS4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NPAS4 - neuronal PAS domain protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318804.1 | 616 | UTR 5 | NP_001305733.1 | |||
NM_178864.3 | 616 | Missense Mutation | TCC,TTC | S,F 147 | NP_849195.2 | |
XM_017017537.1 | 616 | Missense Mutation | TCC,TTC | S,F 224 | XP_016873026.1 | |
XM_017017538.1 | 616 | Intron | XP_016873027.1 | |||
XM_017017539.1 | 616 | Missense Mutation | TCC,TTC | S,F 147 | XP_016873028.1 |