Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAGCGTTGTGGTTTTTTCCCTGG[A/G]CAAATGATTATTTTATAAACAAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603667 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105373738 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105373738 - uncharacterized LOC105373738 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A12 - solute carrier family 25 member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003705.4 | 2366 | UTR 3 | NP_003696.2 | |||
XM_011512070.2 | 2366 | UTR 3 | XP_011510372.1 |