Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTCTAAGCTTTAAGTAAAGAGC[A/T]TGTGACCATTCCTCAATGAGCTCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RBM43 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RBM43 - RNA binding motif protein 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198557.2 | 1460 | Missense Mutation | CAA,CAT | Q,H 285 | NP_940959.1 | |
XM_017004052.1 | 1460 | Missense Mutation | CAA,CAT | Q,H 288 | XP_016859541.1 |