Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCACATACTTTGGTTCATAGCGGA[C/T]GCCTTCTATGTCATGTAAAATGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609803 | ||||||||||||||||||||
Literature Links: |
ANKAR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OSGEPL1 - O-sialoglycoprotein endopeptidase-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022353.2 | 1267 | Missense Mutation | ATC,GTC | I,V 384 | NP_071748.2 | |
XM_005246766.3 | 1267 | Missense Mutation | ATC,GTC | I,V 384 | XP_005246823.1 | |
XM_006712685.1 | 1267 | Missense Mutation | ATC,GTC | I,V 384 | XP_006712748.1 | |
XM_006712686.1 | 1267 | Missense Mutation | ATC,GTC | I,V 237 | XP_006712749.1 | |
XM_011511631.1 | 1267 | Missense Mutation | ATC,GTC | I,V 384 | XP_011509933.1 | |
XM_017004676.1 | 1267 | Missense Mutation | ATC,GTC | I,V 384 | XP_016860165.1 | |
XM_017004677.1 | 1267 | Missense Mutation | ATC,GTC | I,V 384 | XP_016860166.1 | |
XM_017004678.1 | 1267 | Missense Mutation | ATC,GTC | I,V 384 | XP_016860167.1 | |
XM_017004679.1 | 1267 | Missense Mutation | ATC,GTC | I,V 384 | XP_016860168.1 | |
XM_017004680.1 | 1267 | Missense Mutation | ATC,GTC | I,V 384 | XP_016860169.1 | |
XM_017004681.1 | 1267 | Missense Mutation | ATC,GTC | I,V 237 | XP_016860170.1 | |
XM_017004682.1 | 1267 | Missense Mutation | ATC,GTC | I,V 237 | XP_016860171.1 | |
XM_017004683.1 | 1267 | Missense Mutation | ATC,GTC | I,V 237 | XP_016860172.1 | |
XM_017004684.1 | 1267 | Missense Mutation | ATC,GTC | I,V 237 | XP_016860173.1 | |
XM_017004685.1 | 1267 | Missense Mutation | ATC,GTC | I,V 237 | XP_016860174.1 |