Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGATTTTATGGAAAGACTTCATA[C/G]CTTTTTTCTGAGTCAATGTTTCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM117B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM117B - family with sequence similarity 117 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ICA1L - islet cell autoantigen 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288622.1 | 3876 | UTR 3 | NP_001275551.1 | |||
NM_001288623.1 | 3876 | UTR 3 | NP_001275552.1 | |||
NM_001288624.1 | 3876 | Intron | NP_001275553.1 | |||
NM_138468.5 | 3876 | UTR 3 | NP_612477.3 |