Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATTGCTTAAGGATTTGTTTTACT[C/T]TACTACCGTGCCCTAGTTAACACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186760 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD28 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD28 - CD28 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243077.1 | Intron | NP_001230006.1 | ||||
NM_001243078.1 | Intron | NP_001230007.1 | ||||
NM_006139.3 | Intron | NP_006130.1 | ||||
XM_011512194.2 | Intron | XP_011510496.1 | ||||
XM_011512195.2 | Intron | XP_011510497.1 | ||||
XM_011512197.2 | Intron | XP_011510499.1 |