Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGCCAGGAATTAGGTGAGGCAGG[A/G]GCCTGCAAAAGTGACAGATCCTACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600543 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERBB4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERBB4 - erb-b2 receptor tyrosine kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042599.1 | 9822 | UTR 3 | NP_001036064.1 | |||
NM_005235.2 | 9822 | UTR 3 | NP_005226.1 | |||
XM_005246376.2 | 9822 | UTR 3 | XP_005246433.1 | |||
XM_005246377.2 | 9822 | UTR 3 | XP_005246434.1 | |||
XM_006712364.2 | 9822 | UTR 3 | XP_006712427.1 | |||
XM_017003577.1 | 9822 | UTR 3 | XP_016859066.1 | |||
XM_017003578.1 | 9822 | UTR 3 | XP_016859067.1 | |||
XM_017003579.1 | 9822 | UTR 3 | XP_016859068.1 | |||
XM_017003580.1 | 9822 | UTR 3 | XP_016859069.1 | |||
XM_017003581.1 | 9822 | UTR 3 | XP_016859070.1 | |||
XM_017003582.1 | 9822 | UTR 3 | XP_016859071.1 |