Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATAGTAAAAAAGTCAAATTGATC[C/T]TGATAAATTAGAAAAGTAGTGCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616010 MIM: 602676 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COPS7B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COPS7B - COP9 signalosome subunit 7B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282949.2 | Intron | NP_001269878.1 | ||||
NM_001282950.2 | Intron | NP_001269879.1 | ||||
NM_001282951.2 | Intron | NP_001269880.1 | ||||
NM_001282952.2 | Intron | NP_001269881.1 | ||||
NM_001308381.1 | Intron | NP_001295310.1 | ||||
NM_022730.3 | Intron | NP_073567.1 | ||||
XM_006712693.1 | Intron | XP_006712756.1 | ||||
XM_011511638.1 | Intron | XP_011509940.1 | ||||
XM_011511639.1 | Intron | XP_011509941.1 | ||||
XM_011511640.1 | Intron | XP_011509942.1 | ||||
XM_017004716.1 | Intron | XP_016860205.1 | ||||
XM_017004717.1 | Intron | XP_016860206.1 |
PDE6D - phosphodiesterase 6D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |