Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGTCAGTGTCTGATGTGCCACTG[C/T]CACTAGAGTCAGAAGAGCTCTGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610464 | ||||||||||||||||||||
Literature Links: |
GPR156 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR156 - G protein-coupled receptor 156 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168271.1 | 2786 | Missense Mutation | GAC,GGC | D,G 771 | NP_001161743.1 | |
NM_153002.2 | 2786 | Missense Mutation | GAC,GGC | D,G 775 | NP_694547.2 | |
XM_005247165.2 | 2786 | Missense Mutation | GAC,GGC | D,G 775 | XP_005247222.1 | |
XM_011512484.2 | 2786 | Missense Mutation | GAC,GGC | D,G 694 | XP_011510786.1 | |
XM_011512487.1 | 2786 | Missense Mutation | GAC,GGC | D,G 694 | XP_011510789.1 | |
XM_011512488.2 | 2786 | Missense Mutation | GAC,GGC | D,G 389 | XP_011510790.1 | |
XM_017005795.1 | 2786 | Missense Mutation | GAC,GGC | D,G 775 | XP_016861284.1 | |
XM_017005796.1 | 2786 | Missense Mutation | GAC,GGC | D,G 775 | XP_016861285.1 | |
XM_017005797.1 | 2786 | Missense Mutation | GAC,GGC | D,G 771 | XP_016861286.1 |