Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGCATGCAGTCAAGCACTTTGA[C/G]AAGGGAATGCTAAACACACTGCAGA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 615939 | |||||||||||||||||||||||
Literature Links: |
HACD2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
HACD2 - 3-hydroxyacyl-CoA dehydratase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017005855.1 | 1028 | UTR 3 | XP_016861344.1 | |||
XM_017005856.1 | 1028 | Intron | XP_016861345.1 |