Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCACTCTTGCAATTGCTCCTGTC[C/T]GCTTTGACAGGTAAATCTTACTTCC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 114851 MIM: 114852 | |||||||||||||||||||||||
Literature Links: |
CPA3 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CPA3 - carboxypeptidase A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001870.3 | 110 | Missense Mutation | CGC,TGC | R,C 20 | NP_001861.2 |
CPB1 - carboxypeptidase B1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |