Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCATCCTAGATGTGGTGAGTGCTG[A/G]CCTCTCTAGCCCTCCTTGGTGTCCT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 610123 | |||||||||||||||||||||||
Literature Links: |
HTR3E PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
HTR3E - 5-hydroxytryptamine receptor 3E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256613.1 | Intron | NP_001243542.1 | ||||
NM_001256614.1 | Intron | NP_001243543.1 | ||||
NM_182589.2 | Intron | NP_872395.2 | ||||
NM_198313.2 | Intron | NP_938055.1 | ||||
NM_198314.2 | Intron | NP_938056.1 |
HTR3E-AS1 - HTR3E antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |