Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGACATATTGGGTTCAGTTTTATC[G/T]GTGAGTTTGAGGTGAAATAGAGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602850 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFAP99 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFAP99 - cilia and flagella associated protein 99 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF4 - ring finger protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185009.2 | Intron | NP_001171938.1 | ||||
NM_001185010.2 | Intron | NP_001171939.1 | ||||
NM_002938.4 | Intron | NP_002929.1 |