Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACGCAGCAGCAGCTCAACATAAGG[C/T]GGCCAAAAGGAATCACTAATTTTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611502 | ||||||||||||||||||||
Literature Links: |
CENPK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CENPK - centromere protein K | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267038.1 | 1105 | Silent Mutation | CCA,CCG | P,P 240 | NP_001253967.1 | |
NM_022145.4 | 1105 | Silent Mutation | CCA,CCG | P,P 240 | NP_071428.2 | |
XM_011543534.2 | 1105 | Silent Mutation | CCA,CCG | P,P 242 | XP_011541836.1 | |
XM_011543535.2 | 1105 | Silent Mutation | CCA,CCG | P,P 242 | XP_011541837.1 | |
XM_011543536.2 | 1105 | Silent Mutation | CCA,CCG | P,P 240 | XP_011541838.1 | |
XM_011543537.2 | 1105 | Silent Mutation | CCA,CCG | P,P 210 | XP_011541839.1 | |
XM_011543538.2 | 1105 | Silent Mutation | CCA,CCG | P,P 209 | XP_011541840.1 | |
XM_011543539.2 | 1105 | Silent Mutation | CCA,CCG | P,P 207 | XP_011541841.1 | |
XM_011543540.2 | 1105 | Intron | XP_011541842.1 | |||
XM_011543541.2 | 1105 | Silent Mutation | CCA,CCG | P,P 176 | XP_011541843.1 | |
XM_011543542.2 | 1105 | Silent Mutation | CCA,CCG | P,P 176 | XP_011541844.1 | |
XM_017009690.1 | 1105 | Silent Mutation | CCA,CCG | P,P 240 | XP_016865179.1 | |
XM_017009691.1 | 1105 | Silent Mutation | CCA,CCG | P,P 240 | XP_016865180.1 | |
XM_017009692.1 | 1105 | Silent Mutation | CCA,CCG | P,P 240 | XP_016865181.1 | |
XM_017009693.1 | 1105 | Intron | XP_016865182.1 | |||
XM_017009694.1 | 1105 | Silent Mutation | CCA,CCG | P,P 210 | XP_016865183.1 | |
XM_017009695.1 | 1105 | Silent Mutation | CCA,CCG | P,P 208 | XP_016865184.1 | |
XM_017009696.1 | 1105 | Intron | XP_016865185.1 | |||
XM_017009697.1 | 1105 | Silent Mutation | CCA,CCG | P,P 179 | XP_016865186.1 | |
XM_017009698.1 | 1105 | Silent Mutation | CCA,CCG | P,P 177 | XP_016865187.1 | |
XM_017009699.1 | 1105 | Intron | XP_016865188.1 | |||
XM_017009700.1 | 1105 | Silent Mutation | CCA,CCG | P,P 162 | XP_016865189.1 | |
XM_017009701.1 | 1105 | Silent Mutation | CCA,CCG | P,P 135 | XP_016865190.1 |