Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATATTTAGCTAACCCTCTCAAAA[C/T]TGCAGGCCTAGTGAGGTTTAAATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTX3 - metaxin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010891.4 | 6819 | UTR 3 | NP_001010891.4 | |||
NM_001167741.1 | 6819 | UTR 3 | NP_001161213.1 | |||
XM_005248495.3 | 6819 | UTR 3 | XP_005248552.1 | |||
XM_017009440.1 | 6819 | UTR 3 | XP_016864929.1 |