Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCAGGTGTGCAACTCACAGGAG[A/G]GCAGCTCCCACACTCAGGAGATTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616474 | ||||||||||||||||||||
Literature Links: |
PGBD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PGBD1 - piggyBac transposable element derived 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184743.1 | 523 | Missense Mutation | AGC,GGC | S,G 35 | NP_001171672.1 | |
NM_032507.3 | 523 | Missense Mutation | AGC,GGC | S,G 35 | NP_115896.1 | |
XM_006715228.2 | 523 | UTR 5 | XP_006715291.1 | |||
XM_011514942.2 | 523 | UTR 5 | XP_011513244.1 | |||
XM_011514943.2 | 523 | UTR 5 | XP_011513245.1 | |||
XM_011514944.2 | 523 | UTR 5 | XP_011513246.1 | |||
XM_017011361.1 | 523 | Missense Mutation | AGC,GGC | S,G 35 | XP_016866850.1 | |
XM_017011362.1 | 523 | UTR 5 | XP_016866851.1 | |||
XM_017011363.1 | 523 | UTR 5 | XP_016866852.1 |
ZSCAN26 - zinc finger and SCAN domain containing 26 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |