Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCCGCGGGGCCCTTTCGGGGACT[A/G]TCGGCCCGGGGTCTTCTCTGGAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 125647 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DSP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DSP - desmoplakin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNRNP48 - small nuclear ribonucleoprotein U11/U12 subunit 48 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152551.3 | Intron | NP_689764.3 | ||||
XM_011514312.2 | Intron | XP_011512614.1 |