Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTCGCACCTGTGCACTGGAGCTG[C/G]TAACTCCTGTCTGCTGGGGTGAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614964 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELFN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ELFN1 - extracellular leucine rich repeat and fibronectin type III domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128636.2 | Intron | NP_001122108.1 | ||||
XM_006715725.3 | Intron | XP_006715788.1 | ||||
XM_006715726.2 | Intron | XP_006715789.1 | ||||
XM_006715727.3 | Intron | XP_006715790.1 | ||||
XM_011515397.2 | Intron | XP_011513699.1 | ||||
XM_011515398.2 | Intron | XP_011513700.1 | ||||
XM_011515401.2 | Intron | XP_011513703.1 | ||||
XM_017012203.1 | Intron | XP_016867692.1 |