Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATACAGGATTTCATGATGTGCTGG[G/T]CAATGAAAGACCTTCTGCTTACATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604368 | ||||||||||||||||||||
Literature Links: |
GPNMB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPNMB - glycoprotein nmb | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005340.1 | 741 | Missense Mutation | GGC,GTC | G,V 30 | NP_001005340.1 | |
NM_002510.2 | 741 | Missense Mutation | GGC,GTC | G,V 30 | NP_002501.1 | |
XM_005249578.2 | 741 | Missense Mutation | GGC,GTC | G,V 30 | XP_005249635.1 | |
XM_017011676.1 | 741 | Missense Mutation | GGC,GTC | G,V 30 | XP_016867165.1 | |
XM_017011677.1 | 741 | Missense Mutation | GGC,GTC | G,V 30 | XP_016867166.1 | |
XM_017011678.1 | 741 | Missense Mutation | GGC,GTC | G,V 30 | XP_016867167.1 |