Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCAGGGGCTGGGAGCTCTGGCCA[C/T]ACAAGTTTCGAAGACCTACTCCCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610334 MIM: 610385 | ||||||||||||||||||||
Literature Links: |
LOC105375566 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105375566 - uncharacterized LOC105375566 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM176A - transmembrane protein 176A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM176B - transmembrane protein 176B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101311.1 | 715 | Missense Mutation | TAT,TGT | Y,C 233 | NP_001094781.1 | |
NM_001101312.1 | 715 | Missense Mutation | TAT,TGT | Y,C 233 | NP_001094782.1 | |
NM_001101314.1 | 715 | Missense Mutation | TAT,TGT | Y,C 196 | NP_001094784.1 | |
NM_014020.3 | 715 | Missense Mutation | TAT,TGT | Y,C 233 | NP_054739.3 | |
XM_006715933.3 | 715 | Missense Mutation | TAT,TGT | Y,C 196 | XP_006715996.2 |