Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCTCAGTGGGATGAGGGTGAAA[C/G]TGCTATTGCCGGCGGCTCCTGTTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605464 MIM: 612205 | ||||||||||||||||||||
Literature Links: |
ABCB8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ABCB8 - ATP binding cassette subfamily B member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282291.1 | 58 | UTR 5 | NP_001269220.1 | |||
NM_001282292.1 | 58 | UTR 5 | NP_001269221.1 | |||
NM_001282293.1 | 58 | Missense Mutation | CTG,GTG | L,V 5 | NP_001269222.1 | |
NM_007188.4 | 58 | UTR 5 | NP_009119.2 |
ATG9B - autophagy related 9B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |