Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGCATGAGGTCATCCTGTGGGGC[A/G]CTATGACTGTAGTTCCAGTAGCGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRSS37 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRSS37 - protease, serine 37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008270.2 | 1776 | Silent Mutation | AGC,AGT | S,S 94 | NP_001008271.2 | |
NM_001171951.1 | 1776 | Silent Mutation | AGC,AGT | S,S 94 | NP_001165422.1 | |
XM_005249946.4 | 1776 | Silent Mutation | AGC,AGT | S,S 94 | XP_005250003.1 | |
XM_005249947.3 | 1776 | Silent Mutation | AGC,AGT | S,S 94 | XP_005250004.1 | |
XM_011515792.2 | 1776 | Silent Mutation | AGC,AGT | S,S 31 | XP_011514094.1 | |
XM_011515793.2 | 1776 | Silent Mutation | AGC,AGT | S,S 31 | XP_011514095.1 |