Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATACTGGCCTCTTCTCTTTTTCA[G/T]ATGGGGAAACTGAGGCCCAGGGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604318 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTF2IRD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GTF2IRD1 - GTF2I repeat domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199207.1 | Intron | NP_001186136.1 | ||||
NM_005685.3 | Intron | NP_005676.3 | ||||
NM_016328.2 | Intron | NP_057412.1 | ||||
XM_006716182.3 | Intron | XP_006716245.1 | ||||
XM_006716183.3 | Intron | XP_006716246.1 | ||||
XM_011516713.1 | Intron | XP_011515015.1 | ||||
XM_017012802.1 | Intron | XP_016868291.1 | ||||
XM_017012803.1 | Intron | XP_016868292.1 | ||||
XM_017012804.1 | Intron | XP_016868293.1 | ||||
XM_017012805.1 | Intron | XP_016868294.1 |