Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTAGGTCTCTTCAGCAGATCCAG[G/T]ACTGCCGCAGCTCTCTGAACAAGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614511 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MDFIC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MDFIC - MyoD family inhibitor domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166345.1 | 704 | Intron | NP_001159817.1 | |||
NM_001166346.1 | 704 | Missense Mutation | AGG,AGT | R,S 147 | NP_001159818.3 | |
NM_199072.4 | 704 | Intron | NP_951038.1 |