Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGCCCAGAAATGTAGACATGCAA[C/T]TGTATTGACACATCACATAGTTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613138 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSPAN12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TSPAN12 - tetraspanin 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012338.3 | Intron | NP_036470.1 | ||||
XM_005250239.2 | Intron | XP_005250296.1 | ||||
XM_011515993.1 | Intron | XP_011514295.1 | ||||
XM_011515994.1 | Intron | XP_011514296.1 | ||||
XM_017011912.1 | Intron | XP_016867401.1 | ||||
XM_017011913.1 | Intron | XP_016867402.1 | ||||
XM_017011914.1 | Intron | XP_016867403.1 |