Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTGCACTGGCATGTGACGGGTTA[A/G]GTTATAGTGCGCATTAAAGACCTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613301 | ||||||||||||||||||||
Literature Links: |
FEZF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FEZF1 - FEZ family zinc finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024613.3 | 2418 | Silent Mutation | CTA,TTA | L,L 275 | NP_001019784.2 | |
NM_001160264.2 | 2418 | Silent Mutation | CTA,TTA | L,L 225 | NP_001153736.1 | |
XM_005250337.3 | 2418 | Silent Mutation | CTA,TTA | L,L 275 | XP_005250394.1 | |
XM_011516202.2 | 2418 | Silent Mutation | CTA,TTA | L,L 225 | XP_011514504.1 |
FEZF1-AS1 - FEZF1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |