Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAAATTGAAAAAAAGAAAAGAA[C/G]GTGCAAAAGGTAAGACACTTGAGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ANKRD18B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKRD18B - ankyrin repeat domain 18B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244752.1 | 957 | Missense Mutation | NP_001231681.1 | |||
XM_011517881.2 | 957 | Intron | XP_011516183.1 | |||
XM_011517883.2 | 957 | Intron | XP_011516185.1 | |||
XM_011517885.2 | 957 | Intron | XP_011516187.1 | |||
XM_017014728.1 | 957 | Intron | XP_016870217.1 |