Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAACTCTGCTCGGGTCTCAGCCT[C/G]GGGGTAAAGGCAGGATGGGAGAAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603179 MIM: 190990 | ||||||||||||||||||||
Literature Links: |
CA9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CA9 - carbonic anhydrase 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TPM2 - tropomyosin 2 (beta) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301226.1 | Intron | NP_001288155.1 | ||||
NM_001301227.1 | Intron | NP_001288156.1 | ||||
NM_003289.3 | Intron | NP_003280.2 | ||||
NM_213674.1 | Intron | NP_998839.1 | ||||
XM_017015087.1 | Intron | XP_016870576.1 | ||||
XM_017015088.1 | Intron | XP_016870577.1 | ||||
XM_017015089.1 | Intron | XP_016870578.1 | ||||
XM_017015090.1 | Intron | XP_016870579.1 | ||||
XM_017015091.1 | Intron | XP_016870580.1 | ||||
XM_017015092.1 | Intron | XP_016870581.1 | ||||
XM_017015093.1 | Intron | XP_016870582.1 |