Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGCTGATAGGAAAGCTTCGAAGA[A/C]GACAAGATTTGGAGCTCACTCAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MFSD14B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MFSD14B - major facilitator superfamily domain containing 14B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032558.2 | Intron | NP_115947.2 | ||||
XM_006717305.3 | Intron | XP_006717368.1 | ||||
XM_011519114.2 | Intron | XP_011517416.1 | ||||
XM_017015217.1 | Intron | XP_016870706.1 | ||||
XM_017015218.1 | Intron | XP_016870707.1 | ||||
XM_017015219.1 | Intron | XP_016870708.1 | ||||
XM_017015220.1 | Intron | XP_016870709.1 | ||||
XM_017015221.1 | Intron | XP_016870710.1 |