Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAGTCTTTGTACATATTCAAATA[G/T]TTACATGTGGAAAAGATTAAATAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604761 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC01504 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC01504 - long intergenic non-protein coding RNA 1504 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZFAND5 - zinc finger AN1-type containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102420.2 | 4500 | UTR 3 | NP_001095890.1 | |||
NM_001102421.2 | 4500 | UTR 3 | NP_001095891.1 | |||
NM_001278243.1 | 4500 | UTR 3 | NP_001265172.1 | |||
NM_001278244.1 | 4500 | UTR 3 | NP_001265173.1 | |||
NM_001278245.1 | 4500 | UTR 3 | NP_001265174.1 | |||
NM_006007.3 | 4500 | UTR 3 | NP_005998.1 |