Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAACTGTGGAGCCATTTTCCTTGC[A/G]TTGTCCAGAAGGAACTTCGTCCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601296 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MUSK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MUSK - muscle associated receptor tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166280.1 | 673 | UTR 5 | NP_001159752.1 | |||
NM_001166281.1 | 673 | UTR 5 | NP_001159753.1 | |||
NM_005592.3 | 673 | UTR 5 | NP_005583.1 | |||
XM_005251994.3 | 673 | UTR 5 | XP_005252051.1 | |||
XM_005251995.3 | 673 | UTR 5 | XP_005252052.1 | |||
XM_005251996.3 | 673 | UTR 5 | XP_005252053.1 | |||
XM_011518708.2 | 673 | Intron | XP_011517010.1 | |||
XM_017014734.1 | 673 | UTR 5 | XP_016870223.1 |