Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCATGAAGCTGCCAACTTCATAC[C/T]CATCGGGGACCTCTCTGGGCAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605416 MIM: 614460 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DKK3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DKK3 - dickkopf WNT signaling pathway inhibitor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018057.1 | 972 | Missense Mutation | GAG,GGG | E,G 307 | NP_001018067.1 | |
NM_013253.4 | 972 | Missense Mutation | GAG,GGG | E,G 307 | NP_037385.2 | |
NM_015881.5 | 972 | Missense Mutation | GAG,GGG | E,G 307 | NP_056965.3 | |
XM_006718178.2 | 972 | Missense Mutation | GAG,GGG | E,G 307 | XP_006718241.1 | |
XM_017017553.1 | 972 | Missense Mutation | GAG,GGG | E,G 321 | XP_016873042.1 | |
XM_017017554.1 | 972 | Missense Mutation | GAG,GGG | E,G 321 | XP_016873043.1 | |
XM_017017555.1 | 972 | Missense Mutation | GAG,GGG | E,G 321 | XP_016873044.1 |
USP47 - ubiquitin specific peptidase 47 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |