Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATATACAATTTAGAAAAATAATCT[C/T]ATTCATGACATCTCTGTGTGAGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612815 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZDHHC13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZDHHC13 - zinc finger DHHC-type containing 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001483.2 | Intron | NP_001001483.1 | ||||
NM_019028.2 | Intron | NP_061901.2 | ||||
XM_005252996.2 | Intron | XP_005253053.1 | ||||
XM_011520193.1 | Intron | XP_011518495.1 | ||||
XM_011520194.2 | Intron | XP_011518496.1 | ||||
XM_011520195.1 | Intron | XP_011518497.1 |