Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGAAAAAGAAAAGTATGAATCAA[A/G]TGGAATGGAAGAATTTCTACACAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600367 MIM: 612296 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSTF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSTF3 - cleavage stimulation factor subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033505.1 | Intron | NP_001028677.1 | ||||
NM_001033506.1 | Intron | NP_001028678.1 | ||||
NM_001326.2 | Intron | NP_001317.1 |
LINC00294 - long intergenic non-protein coding RNA 294 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TCP11L1 - t-complex 11 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |