Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTTTTTTTTTTGGTTGTTGTTA[A/C]ACAGAACCTTAAGTTTATGTTTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608858 MIM: 605181 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGFR1OP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FGFR1OP2 - FGFR1 oncogene partner 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171887.1 | 2837 | UTR 3 | NP_001165358.1 | |||
NM_001171888.1 | 2837 | Intron | NP_001165359.1 | |||
NM_015633.2 | 2837 | UTR 3 | NP_056448.1 |
TM7SF3 - transmembrane 7 superfamily member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016551.2 | 2837 | Intron | NP_057635.1 | |||
XM_005253391.3 | 2837 | UTR 3 | XP_005253448.1 | |||
XM_011520690.2 | 2837 | UTR 3 | XP_011518992.1 | |||
XM_011520691.1 | 2837 | Intron | XP_011518993.1 | |||
XM_017019463.1 | 2837 | UTR 3 | XP_016874952.1 | |||
XM_017019464.1 | 2837 | UTR 3 | XP_016874953.1 | |||
XM_017019465.1 | 2837 | UTR 3 | XP_016874954.1 | |||
XM_017019466.1 | 2837 | UTR 3 | XP_016874955.1 |