Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGACTGCTTTCAGGCCACATACCT[C/T]GGCCTTGTTGTTGAAAACATGTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606222 MIM: 609388 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGSF6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGSF6 - immunoglobulin superfamily member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005849.3 | Intron | NP_005840.2 |
METTL9 - methyltransferase like 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077180.2 | Intron | NP_001070648.1 | ||||
NM_001288659.1 | Intron | NP_001275588.1 | ||||
NM_001288660.1 | Intron | NP_001275589.1 | ||||
NM_016025.4 | Intron | NP_057109.3 | ||||
XM_017023269.1 | Intron | XP_016878758.1 |