Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTCCTGTCACTTCCTGTGACTC[A/G]GGTACAGCTCCATGGGAATGTGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLYWCH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FLYWCH1 - FLYWCH-type zinc finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308068.1 | 491 | Intron | NP_001294997.1 | |||
NM_020912.1 | 491 | Intron | NP_065963.1 | |||
NM_032296.2 | 491 | Intron | NP_115672.2 | |||
XM_006720962.3 | 491 | Intron | XP_006721025.1 | |||
XM_011522701.2 | 491 | Intron | XP_011521003.1 | |||
XM_011522702.2 | 491 | Intron | XP_011521004.1 | |||
XM_011522703.2 | 491 | Intron | XP_011521005.1 | |||
XM_011522704.2 | 491 | Intron | XP_011521006.1 | |||
XM_011522705.2 | 491 | Intron | XP_011521007.1 | |||
XM_011522707.2 | 491 | Intron | XP_011521009.1 | |||
XM_017023772.1 | 491 | UTR 5 | XP_016879261.1 |