Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACGTGATCCAGCGCTTTGAGAGGC[A/G]GGGCTTCACGCTGGTGGGGATGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615839 MIM: 601818 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DECR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DECR2 - 2,4-dienoyl-CoA reductase 2, peroxisomal | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100134368 - uncharacterized LOC100134368 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NME4 - NME/NM23 nucleoside diphosphate kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286433.1 | 222 | Missense Mutation | CAG,CGG | Q,R 64 | NP_001273362.1 | |
NM_001286435.1 | 222 | Missense Mutation | CAG,CGG | Q,R 64 | NP_001273364.1 | |
NM_001286436.1 | 222 | UTR 5 | NP_001273365.1 | |||
NM_001286438.1 | 222 | UTR 5 | NP_001273367.1 | |||
NM_001286439.1 | 222 | UTR 5 | NP_001273368.1 | |||
NM_001286440.1 | 222 | UTR 5 | NP_001273369.1 | |||
NM_005009.2 | 222 | Missense Mutation | CAG,CGG | Q,R 64 | NP_005000.1 |