Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGGGCTGAGCTCTAGGCAGGGA[C/T]GCTCCAGGAAGAAGTGGGTCACCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611026 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FA2H PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FA2H - fatty acid 2-hydroxylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024306.4 | 1137 | UTR 3 | NP_077282.3 | |||
XM_011523317.2 | 1137 | Intron | XP_011521619.1 | |||
XM_011523319.2 | 1137 | UTR 3 | XP_011521621.1 |