Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGGAGCTTGTCCACCAGGTGCAC[C/T]GGCGTGCAGGACACGACCGCCCGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609226 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB40B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAB40B - RAB40B, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006822.2 | 853 | Silent Mutation | CCA,CCG | P,P 205 | NP_006813.1 | |
XM_005256334.4 | 853 | Silent Mutation | CCA,CCG | P,P 145 | XP_005256391.1 | |
XM_005256335.3 | 853 | Intron | XP_005256392.1 | |||
XM_006722271.3 | 853 | Silent Mutation | CCA,CCG | P,P 165 | XP_006722334.1 | |
XM_011523528.2 | 853 | Silent Mutation | CCA,CCG | P,P 188 | XP_011521830.1 | |
XM_017024042.1 | 853 | Intron | XP_016879531.1 | |||
XM_017024043.1 | 853 | Silent Mutation | CCA,CCG | P,P 145 | XP_016879532.1 |
WDR45B - WD repeat domain 45B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |