Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCTCGGTCGGCAAGGAAATCCAT[C/T]AGGTGGTCATATAAGGCCTGCAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601269 MIM: 614405 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1QBP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1QBP - complement C1q binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001212.3 | Intron | NP_001203.1 |
DHX33 - DEAH-box helicase 33 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPAIN - RPA interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033002.3 | Intron | NP_001028174.2 | ||||
NM_001160243.1 | Intron | NP_001153715.1 | ||||
NM_001160244.1 | Intron | NP_001153716.1 | ||||
NM_001160246.1 | Intron | NP_001153718.1 | ||||
NM_001160266.1 | Intron | NP_001153738.1 |